2-Bromo-4-(trifluoromethoxy)benzonitrile ≥95%
Supplier: Matrix Scientific
074675-250MG
074675-1G
074675-5G
102537-640EA
2267.69
USD
102537-640
102537-642
102537-644
2-Bromo-4-(trifluoromethoxy)benzonitrile ≥95%
2-Bromo-4-(trifluoromethoxy)benzonitrile
Hazard: Toxic
Formula:
C₈H₃BrF₃NO |
MDL Number:
MFCD13185874 CAS Number: 1214334-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...