Dibenzothiophene-2-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Dibenzo[b,d]thiophen-2-ylboronic acid

074673-1G 074673-5G
102537-632EA 463.08 USD
102537-632 102537-634
Dibenzothiophene-2-boronic acid ≥95%
Dibenzothiophene-2-boronic acid

Hazard: Irritant

Formula: C₁₂H₉BO₂S
MW: 228.07 g/mol
MDL Number: MFCD01318982
CAS Number: 668983-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR