1-Methyl-1H-pyrazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

074671-1G 074671-5G
102537-624EA 75.38 USD
102537-624 102537-626
1-Methyl-1H-pyrazole-3-carbaldehyde ≥95%
1-Methyl-1H-pyrazole-3-carbaldehyde

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.1100 g/mol
Boiling Pt: 88-90°C/0.1mm
MDL Number: MFCD03419800
CAS Number: 27258-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR