3-Fluoro-5-methoxybenzyl bromide ≥95%

Supplier: Matrix Scientific

074670-1G 074670-5G
102537-620EA 533.85 USD
102537-620 102537-622
3-Fluoro-5-methoxybenzyl bromide ≥95%
3-Fluoro-5-methoxybenzyl bromide

Hazard: Irritant

Formula: C₈H₈BrFO
MW: 219.0509 g/mol
MDL Number: MFCD08741378
CAS Number: 914637-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR