1-Bromo-3-chloro-5-iodobenzene ≥95%

Supplier: Matrix Scientific

074668-1G 074668-5G 074668-25G
102537-608EA 44.62 USD
102537-608 102537-610 102551-910
1-Bromo-3-chloro-5-iodobenzene ≥95%
1-Bromo-3-chloro-5-iodobenzene

Hazard: Irritant

Formula: C₆H₃BrClI
MDL Number: MFCD07778998
CAS Number: 13101-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR