4-Bromo-2-chlorobenzo[d]thiazole ≥95%

Supplier: Matrix Scientific

074665-1G 074665-5G
102537-594EA 646.15 USD
102537-594 102537-596
4-Bromo-2-chlorobenzo[d]thiazole ≥95%
4-Bromo-2-chlorobenzo[d]thiazole

Hazard: Irritant

Formula: C₇H₃BrClNS
MDL Number: MFCD04971817
CAS Number: 182344-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR