Methyl-4-fluoro-3-(trifluoromethyl)benzoate ≥95%

Supplier: Matrix Scientific

074664-1G 074664-5G
102537-590EA 56.92 USD
102537-590 102537-592
Methyl-4-fluoro-3-(trifluoromethyl)benzoate ≥95%
Methyl-4-fluoro-3-(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₉H₆F₄O₂
MDL Number: MFCD06203709
CAS Number: 176694-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR