(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected
074662-1G
074662-5G
102537-578EA
60
USD
102537-578
102537-580
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid ≥95%
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
Hazard: Irritant/Store in Freezer
Formula:
C₈H₁₃BN₂O₄ |
MDL Number:
MFCD09951915 CAS Number: 1188405-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...