Dimethyl-2-(2-methoxyphenoxy)malonate ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-(2-methoxyphenoxy)malonate

074660-1G 074660-5G 074660-10G
102537-566EA 213.85 USD
102537-566 102537-568 102537-570
Dimethyl-2-(2-methoxyphenoxy)malonate ≥95%
Dimethyl-2-(2-methoxyphenoxy)malonate

Hazard: Irritant

Formula: C₁₂H₁₄O₆
MW: 254.2360 g/mol
MDL Number: MFCD06797652
CAS Number: 150726-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR