1,3-Dimethyl-1H-pyrazol-5-ol ≥95%

Supplier: Matrix Scientific

074659-1G 074659-5G 074659-25G
102537-560EA 90.77 USD
102537-560 102537-562 102833-706
1,3-Dimethyl-1H-pyrazol-5-ol ≥95%
1,3-Dimethyl-1H-pyrazol-5-ol

Hazard: Irritant

Formula: C₅H₈N₂O
MW: 112.1300 g/mol
MDL Number: MFCD00129673
CAS Number: 5203-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR