5-Bromo-2-chlorobenzothiazole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-chlorobenzo[d]thiazole
074656-1G
074656-5G
102537-546EA
307.69
USD
102537-546
102537-548
5-Bromo-2-chlorobenzothiazole ≥95%
5-Bromo-2-chlorobenzothiazole
Hazard: Irritant
Formula:
C₇H₃BrClNS MW: 248.53 g/mol |
MDL Number:
MFCD09749227 CAS Number: 824403-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...