2-(4-Aminophenyl)-2-methylpropanenitrile ≥95%
Supplier: Matrix Scientific
074655-1G
074655-5G
102537-540EA
640
USD
102537-540
102537-542
2-(4-Aminophenyl)-2-methylpropanenitrile ≥95%
2-(4-Aminophenyl)-2-methylpropanenitrile
Hazard: Irritant
Formula:
C₁₀H₁₂N₂ MW: 160.2200 g/mol |
MDL Number:
MFCD09909912 CAS Number: 115279-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...