7-Chloroquinoline ≥95%

Supplier: Matrix Scientific

074653-1G 074653-5G 074653-10G
102537-528EA 355.38 USD
102537-528 102537-530 102537-532
7-Chloroquinoline ≥95%
7-Chloroquinoline

Hazard: Irritant

Formula: C₉H₆ClN
MDL Number: MFCD00956370
CAS Number: 612-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR