Methyl-5-quinolyl ether ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Methoxyquinoline
074651-1G
074651-5G
102537-520EA
70.77
USD
102537-520
102537-522
Methyl-5-quinolyl ether ≥95%
Methyl 5-quinolyl ether
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.1900 g/mol |
MDL Number:
MFCD02752664 CAS Number: 6931-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...