4-(4-Chlorophenoxy)phenol ≥95%
Supplier: Matrix Scientific
074649-1G
074649-5G
074649-10G
102537-510EA
90.77
USD
102537-510
102537-512
102537-514
4-(4-Chlorophenoxy)phenol ≥95%
4-(4-Chlorophenoxy)phenol
Hazard: Irritant
Formula:
C₁₂H₉ClO₂ |
MDL Number:
MFCD00125839 CAS Number: 21567-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...