3-Chloroquinoline ≥95%

Supplier: Matrix Scientific

074648-1G 074648-5G
102537-506EA 126.15 USD
102537-506 102537-508
3-Chloroquinoline ≥95%
3-Chloroquinoline

Hazard: Irritant

Formula: C9H6ClN
MW: 163.6036 g/mol
MDL Number: MFCD00234489
CAS Number: 612-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR