Methyl-3-quinolyl ether ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methoxyquinoline
074647-1G
074647-5G
102537-502EA
346.15
USD
102537-502
102537-504
Methyl-3-quinolyl ether ≥95%
Methyl 3-quinolyl ether
Hazard: Irritant
Formula:
C₁₀H₉NO |
MDL Number:
MFCD16619425 CAS Number: 6931-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...