2-Methylnicotinamide ≥95%

Supplier: Matrix Scientific

074646-5G 074646-25G
102537-498EA 113.85 USD
102537-498 102833-704
2-Methylnicotinamide ≥95%
2-Methylnicotinamide

Hazard: Irritant

Formula: C₇H₈N₂O
MDL Number: MFCD09909457
CAS Number: 58539-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR