5-Bromo-2-ethoxypyridin-3-amine ≥95%

Supplier: Matrix Scientific

074644-1G 074644-5G
102537-486EA 173.85 USD
102537-486 102537-488
5-Bromo-2-ethoxypyridin-3-amine ≥95%
5-Bromo-2-ethoxypyridin-3-amine

Hazard: Irritant

Formula: C₇H₉BrN₂O
MDL Number: MFCD07375021
CAS Number: 886373-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR