5-(2-Methyl-1H-imidazol-1-yl)-2-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
074642-250MG
074642-1G
074642-5G
102537-474EA
355.38
USD
102537-474
102537-476
102537-478
5-(2-Methyl-1H-imidazol-1-yl)-2-nitrobenzoic acid ≥95%
5-(2-Methyl-1H-imidazol-1-yl)-2-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₁H₉N₃O₄ MW: 247.21 g/mol |
MDL Number:
MFCD09738922 CAS Number: 954265-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...