3-Amino-2-methoxy-4-picoline ≥95%

Supplier: Matrix Scientific

074639-1G 074639-5G
102537-460EA 240 USD
102537-460 102537-462
3-Amino-2-methoxy-4-picoline ≥95%
3-Amino-2-methoxy-4-picoline

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD08277287
CAS Number: 76005-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR