6-Chloro-2-ethoxypyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

074638-250MG 074638-1G
102537-454EA 826.15 USD
102537-454 102537-456
6-Chloro-2-ethoxypyrimidin-4-amine ≥95%
6-Chloro-2-ethoxypyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₈ClN₃O
MW: 173.6 g/mol
MDL Number: MFCD18205868
CAS Number: 3286-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR