3-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥95%

Supplier: Matrix Scientific

074636-250MG 074636-1G 074636-5G
102537-442EA 640 USD
102537-442 102537-444 102537-446
3-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥95%
3-Bromo[1,2,4]triazolo[4,3-a]pyridine

Hazard: Irritant

Formula: C6H4BrN3
MW: 198.0200 g/mol
MDL Number: MFCD11111699
CAS Number: 4922-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR