Ethyl quinoline-6-carboxylate ≥95%
Supplier: Matrix Scientific
074634-1G
074634-5G
074634-10G
102537-432EA
355.38
USD
102537-432
102537-434
102537-436
Ethyl quinoline-6-carboxylate ≥95%
Ethyl quinoline-6-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁NO₂ MW: 201.2200 g/mol |
MDL Number:
MFCD00047612 CAS Number: 73987-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...