tert-Butyl-2-amino-4-fluorophenylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (2-Amino-4-fluorophenyl)carbamate, N1-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl 2-amino-4-fluorophenylcarbamate, (2-Amino-4-fluorophenyl)carbamic acid tert-Butyl ester, N1-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine

074633-250MG 074633-1G 074633-5G
102537-426EA 400 USD
102537-426 102537-428 102537-430
tert-Butyl-2-amino-4-fluorophenylcarbamate ≥95%
tert-Butyl-2-amino-4-fluorophenylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O₂
MDL Number: MFCD09701246
CAS Number: 579474-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR