N-Methyl-4-(methylsulfonyl)aniline hydrochloride ≥95%

Supplier: Matrix Scientific

074631-250MG 074631-1G 074631-5G
102537-414EA 400 USD
102537-414 102537-416 102537-418
N-Methyl-4-(methylsulfonyl)aniline hydrochloride ≥95%
N-Methyl-4-(methylsulfonyl)aniline hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO₂S
MW: 221.7000 g/mol
MDL Number: MFCD18157693
CAS Number: 1263378-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR