3-Amino-6-chloropyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074627-1G 074627-5G
102537-396EA 640 USD
102537-396 102537-398
3-Amino-6-chloropyridine-2-carboxylic acid ≥95%
3-Amino-6-chloropyridine-2-carboxylic acid

Hazard: Irritant

Formula: C6H5ClN2O2
MW: 172.5691 g/mol
MDL Number: MFCD09264291
CAS Number: 866807-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR