[1,2,4]Triazolo[4,3-a]pyrazine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

074624-250MG 074624-1G
102537-382EA 1110.77 USD
102537-382 102537-384
[1,2,4]Triazolo[4,3-a]pyrazine-3-carboxylic acid ≥95%
[1,2,4]Triazolo[4,3-a]pyrazine-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₄N₄O₂
CAS Number: 1245644-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR