N-Boc-D-prolinal ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
074620-1G
074620-5G
074620-10G
102537-362EA
133.85
USD
102537-362
102537-364
102537-366
N-Boc-D-prolinal ≥95%
N-Boc-D-prolinal
Hazard: Irritant
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol |
MDL Number:
MFCD01321389 CAS Number: 73365-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...