3-Bromo-6-chloro-7-azaindole ≥95%

Supplier: Matrix Scientific

074617-250MG 074617-1G
102537-346EA 640 USD
102537-346 102537-348
3-Bromo-6-chloro-7-azaindole ≥95%
3-Bromo-6-chloro-7-azaindole

Hazard: Irritant

Formula: C7H4BrClN2
MW: 231.4771 g/mol
MDL Number: MFCD12962923
CAS Number: 1190321-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR