5-Bromo-2-ethylpyrimidine ≥95%

Supplier: Matrix Scientific

074615-250MG 074615-1G
102537-338EA 670.77 USD
102537-338 102537-340
5-Bromo-2-ethylpyrimidine ≥95%
5-Bromo-2-ethylpyrimidine

Hazard: Irritant

Formula: C₆H₇BrN₂
CAS Number: 873331-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR