4-Fluoro-3-nitrotoluene ≥95%

Supplier: Matrix Scientific

074614-10G 074614-100G
102537-334EA 30.77 USD
102537-334 102537-336
4-Fluoro-3-nitrotoluene ≥95%
4-Fluoro-3-nitrotoluene

Hazard: Irritant

Formula: C₇H₆FNO₂
MDL Number: MFCD00007060
CAS Number: 446-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR