4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

074612-250MG 074612-1G 074612-5G
102537-322EA 1515.38 USD
102537-322 102537-324 102537-326
4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine ≥95%
4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₆H₄Cl₂N₄
MW: 203.0288 g/mol
MDL Number: MFCD16249844
CAS Number: 1211522-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR