3-Bromopyridazine ≥95%
Supplier: Matrix Scientific
074611-1G
074611-5G
102537-316EA
150.77
USD
102537-316
102537-318
3-Bromopyridazine ≥95%
3-Bromopyridazine
Hazard: Irritant/Store in Freezer
Formula:
C₄H₃BrN₂ MW: 158.9840 g/mol Boiling Pt: 270°C Density: 1.727 Flash Pt: 117°C |
MDL Number:
MFCD10688587 CAS Number: 88491-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...