4-Chloro-1H-imidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1H-imidazole

074610-1G 074610-5G
102537-312EA 136.92 USD
102537-312 102537-314
4-Chloro-1H-imidazole ≥95%
4-Chloro-1H-imidazole

Hazard: Irritant

Formula: C₃H₃ClN₂
MW: 102.52 g/mol
MDL Number: MFCD09033087
CAS Number: 15965-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR