3-(4-Fluorophenyl)phenethyl alcohol ≥95%
Supplier: Matrix Scientific
074609-250MG
074609-1G
074609-5G
102537-304EA
740
USD
102537-304
102537-306
102537-308
3-(4-Fluorophenyl)phenethyl alcohol ≥95%
3-(4-Fluorophenyl)phenethyl alcohol
Hazard: Irritant
Formula:
C₁₄H₁₃FO MW: 216.25 g/mol |
MDL Number:
MFCD06201087 CAS Number: 840521-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...