3-(3-Fluorophenyl)phenethyl alcohol ≥95%

Supplier: Matrix Scientific

074608-250MG 074608-1G 074608-5G
102537-298EA 740 USD
102537-298 102537-300 102537-302
3-(3-Fluorophenyl)phenethyl alcohol ≥95%
3-(3-Fluorophenyl)phenethyl alcohol

Hazard: Irritant

Formula: C₁₄H₁₃FO
CAS Number: 840522-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR