3-Bromo-6-methylpyridazine ≥95%

Supplier: Matrix Scientific

074604-1G 074604-5G
102537-280EA 167.69 USD
102537-280 102537-282
3-Bromo-6-methylpyridazine ≥95%
3-Bromo-6-methylpyridazine

Hazard: Irritant

Formula: C5H5BrN2
MW: 173.0106 g/mol
MDL Number: MFCD09831907
CAS Number: 65202-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR