N-(2-Hydroxyethyl)nicotinamide ≥95%

Supplier: Matrix Scientific

074603-250MG 074603-1G 074603-5G
102537-274EA 427.69 USD
102537-274 102537-276 102537-278
N-(2-Hydroxyethyl)nicotinamide ≥95%
N-(2-Hydroxyethyl)nicotinamide

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD00547113
CAS Number: 6265-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR