4-Bromo-2,6-dimethylanisole ≥95%

Supplier: Matrix Scientific

074601-10G 074601-100G
102537-268EA 30.77 USD
102537-268 102537-270
4-Bromo-2,6-dimethylanisole ≥95%
4-Bromo-2,6-dimethylanisole

Hazard: Irritant

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 117-120°C
MDL Number: MFCD00143259
CAS Number: 14804-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR