N-Methylpiperidin-3-amine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Methylamino)piperidine dihydrochloride
074600-1G
074600-5G
074600-10G
102537-262EA
178.46
USD
102537-262
102537-264
102537-266
N-Methylpiperidin-3-amine dihydrochloride ≥95%
N-Methylpiperidin-3-amine dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₆Cl₂N₂ MW: 187.1100 g/mol |
MDL Number:
MFCD00673143 CAS Number: 127294-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...