Methyl-4-chloronicotinate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-chloronicotinate hydrochloride

074599-250MG 074599-1G 074599-5G
102537-256EA 355.38 USD
102537-256 102537-258 102537-260
Methyl-4-chloronicotinate hydrochloride ≥95%
Methyl-4-chloronicotinate hydrochloride

Hazard: Irritant

Formula: C₇H₇Cl₂NO₂
MW: 208.0400 g/mol
MDL Number: MFCD18157697
CAS Number: 1351479-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR