3-Chloro-6-(methylsulfonyl)pyridazine ≥95%

Supplier: Matrix Scientific

074597-1G 074597-5G
102537-246EA 640 USD
102537-246 102537-248
3-Chloro-6-(methylsulfonyl)pyridazine ≥95%
3-Chloro-6-(methylsulfonyl)pyridazine

Hazard: Irritant

Formula: C5H5ClN2O2S
MDL Number: MFCD03931474
CAS Number: 7145-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR