(5-Bromo-2-chlorophenyl)methanamine ≥95%

Supplier: Matrix Scientific

074595-1G 074595-5G 074595-10G
102537-234EA 533.85 USD
102537-234 102537-236 102537-238
(5-Bromo-2-chlorophenyl)methanamine ≥95%
(5-Bromo-2-chlorophenyl)methanamine

Hazard: Irritant

Formula: C₇H₇BrClN
MW: 220.4900 g/mol
MDL Number: MFCD12026286
CAS Number: 1096296-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR