1-Boc-4-(3-aminopyridin-2-yl)piperazine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(3-Aminopyridin-2-yl)piperazine
074584-1G
074584-10G
102537-176EA
533.85
USD
102537-176
102537-180
1-Boc-4-(3-aminopyridin-2-yl)piperazine ≥95%
1-Boc-4-(3-aminopyridin-2-yl)piperazine
Hazard: Irritant
Formula:
C₁₄H₂₂N₄O₂ MW: 278.3501 g/mol |
MDL Number:
MFCD03407996 CAS Number: 111669-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...