tert-Butyl (2-aminophenyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, 2-(tert-Butoxycarbonylamino)aniline, (2-Aminophenyl)carbamic acid tert-Butyl ester, 2-(Boc-amino)aniline, N-Boc-1,2-phenylenediamine

074576-1G 074576-10G
102537-134EA 144.62 USD
102537-134 102537-138
tert-Butyl (2-aminophenyl)carbamate ≥95%
tert-Butyl (2-aminophenyl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.2600 g/mol
MDL Number: MFCD02169707
CAS Number: 146651-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR