3-Bromo-5-fluoro-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

074569-5G 074569-25G
102537-100EA 120 USD
102537-100 102833-698
3-Bromo-5-fluoro-2-methoxypyridine ≥95%
3-Bromo-5-fluoro-2-methoxypyridine

Hazard: Irritant/Store in Freezer

Formula: C₆H₅BrFNO
MDL Number: MFCD04972397
CAS Number: 884494-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR