Cis-4-Hydroxy-D-proline hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(2R,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride, H-cis-D-Hyp-OH·HCl
074556-5G
074556-25G
102537-036EA
186.15
USD
102537-036
102551-900
Cis-4-Hydroxy-D-proline hydrochloride ≥95%
Cis-4-Hydroxy-D-proline hydrochloride
Hazard: Irritant
Formula:
C₅H₁₀ClNO₃ MW: 167.5908 g/mol |
MDL Number:
MFCD09749867 CAS Number: 77449-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...