2,4-Dichloro-5,6,7,8-tetrahydroquinazoline ≥95%

Supplier: Matrix Scientific

074540-250MG 074540-5G
102536-954EA 138.46 USD
102536-954 102536-958
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline ≥95%
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline

Hazard: Irritant

Formula: C₈H₈Cl₂N₂
MW: 203.0700 g/mol
MDL Number: MFCD12400774
CAS Number: 1127-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR