Methyl-2-(2-formylphenyl)acetate ≥90%

Supplier: Matrix Scientific

074525-250MG 074525-5G
102536-878EA 461.54 USD
102536-878 102536-882
Methyl-2-(2-formylphenyl)acetate ≥90%
Methyl-2-(2-formylphenyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD12755820
CAS Number: 63969-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR